ENA196830150 Enamine

2-(oxolan-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD01029198
  • Cas Number : 86932-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44220
Availability details for
Quantity
  • Availability
  • Add to Cart