ENA196830151 Enamine

2-(trifluoromethyl)-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD01309785
  • Cas Number : 82791-93-3
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44222
Availability details for
Quantity
  • Availability
  • Add to Cart