ENA196830157 Enamine

2-(2-bromophenoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09946847
  • Cas Number : 1016873-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44238
Availability details for
Quantity
  • Availability
  • Add to Cart