ENA196830170 Enamine

6-(2-bromophenoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09937687
  • Cas Number : 1017031-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44272
Availability details for
Quantity
  • Availability
  • Add to Cart