ENA196830174 Enamine

4-[2-(diethylamino)ethoxy]benzoic acid hydrochloride

  • MDL number MFCD12197102
  • Cas Number : 59931-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44282
Availability details for
Quantity
  • Availability
  • Add to Cart