ENA196830184 Enamine

2-ethoxycyclopropane-1-carboxylic acid

  • MDL number MFCD11858184
  • Cas Number : 53381-05-8
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43013

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-43013
pictograms GHS05,GHS07
hstatements H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart