ENA196830188 Enamine

6-chloro-N-(prop-2-yn-1-yl)pyridine-3-carboxamide

  • MDL number MFCD09944097
  • Cas Number : 1017047-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44829
Availability details for
Quantity
  • Availability
  • Add to Cart