ENA196830195 Enamine

methyl 2,4-dichloro-1,3-thiazole-5-carboxylate

  • MDL number MFCD09878042
  • Cas Number : 861708-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43225

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-43225
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart