ENA196830196 Enamine

2-(pyrimidin-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD12093744
  • Cas Number : 1014631-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44853
Availability details for
Quantity
  • Availability
  • Add to Cart