ENA196830197 Enamine

2-[2-(pyrimidin-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD12093745
  • Cas Number : 1154009-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44855
Availability details for
Quantity
  • Availability
  • Add to Cart