ENA196830201 Enamine

2-(3,4-dichlorophenoxy)propanoic acid

  • MDL number MFCD03422223
  • Cas Number : 3307-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44869
Availability details for
Quantity
  • Availability
  • Add to Cart