ENA196830202 Enamine

isocyanatocyclobutane

  • MDL number MFCD09938339
  • Cas Number : 5811-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-44871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44871
Availability details for
Quantity
  • Availability
  • Add to Cart