ENA196830211 Enamine

1-[(pyridin-4-yl)methyl]piperidin-4-amine

  • MDL number MFCD06739068
  • Cas Number : 160357-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44904
Availability details for
Quantity
  • Availability
  • Add to Cart