ENA196830213 Enamine

2-methyl-3-(4-methylpiperazin-1-yl)propan-1-amine

  • MDL number MFCD09901920
  • Cas Number : 50846-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44908
Availability details for
Quantity
  • Availability
  • Add to Cart