ENA196830217 Enamine

N-(5-methylpyridin-2-yl)piperidine-3-carboxamide

  • MDL number MFCD08272024
  • Cas Number : 883106-75-0
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-44924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44924
Availability details for
Quantity
  • Availability
  • Add to Cart