ENA196830222 Enamine

3-(2,3-dihydro-1-benzofuran-2-amido)benzoic acid

  • MDL number MFCD09815165
  • Cas Number : 1016494-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44936
Availability details for
Quantity
  • Availability
  • Add to Cart