ENA196830224 Enamine

N1-ethylbenzene-1,3-diamine

  • MDL number MFCD11132710
  • Cas Number : 50617-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44943
Availability details for
Quantity
  • Availability
  • Add to Cart