ENA196830228 Enamine

methyl[(2-methyl-1,3-thiazol-5-yl)methyl]amine

  • MDL number MFCD02853609
  • Cas Number : 63139-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44959
Availability details for
Quantity
  • Availability
  • Add to Cart