ENA196830229 Enamine

ethyl 3,6-dimethyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylate

  • MDL number MFCD12197134
  • Cas Number : 1181458-53-6
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44962
Availability details for
Quantity
  • Availability
  • Add to Cart