ENA196830232 Enamine

2-[(4-chlorophenyl)amino]acetic acid

  • MDL number MFCD00462901
  • Cas Number : 5465-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44970

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44970
Availability details for
Quantity
  • Availability
  • Add to Cart