ENA196830234 Enamine

N-(5-methylpyridin-2-yl)piperidine-4-carboxamide

  • MDL number MFCD08272005
  • Cas Number : 110105-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44976
Availability details for
Quantity
  • Availability
  • Add to Cart