ENA196830238 Enamine

1-(pyridin-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD09054737
  • Cas Number : 896717-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43488
Availability details for
Quantity
  • Availability
  • Add to Cart