ENA196830239 Enamine

2-(4-methoxyphenyl)oxirane

  • MDL number MFCD06808790
  • Cas Number : 6388-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20-25
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-43519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43519
Availability details for
Quantity
  • Availability
  • Add to Cart