ENA196830241 Enamine

2-(3,4-difluorophenyl)-2-(ethylamino)acetic acid

  • MDL number MFCD12048085
  • Cas Number : 1214017-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45005
Availability details for
Quantity
  • Availability
  • Add to Cart