ENA196830254 Enamine

2-chloro-N-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)propanamide

  • MDL number MFCD09927646
  • Cas Number : 1016686-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28043
Availability details for
Quantity
  • Availability
  • Add to Cart