ENA196830261 Enamine

5-amino-3-(3-hydroxypropoxy)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD09702276
  • Cas Number : 892143-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45103
Availability details for
Quantity
  • Availability
  • Add to Cart