ENA196830266 Enamine

{bicyclo[4.2.0]octa-1(6),2,4-trien-7-yl}methanamine

  • MDL number MFCD06213241
  • Cas Number : 1005-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45115
Availability details for
Quantity
  • Availability
  • Add to Cart