ENA196830271 Enamine

tert-butyl N-(2-amino-4-fluorophenyl)carbamate

  • MDL number MFCD09701246
  • Cas Number : 579474-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45138

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45138
Availability details for
Quantity
  • Availability
  • Add to Cart