ENA196830273 Enamine

2-(cyclopentylamino)acetic acid

  • MDL number MFCD08442382
  • Cas Number : 58988-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45142
Availability details for
Quantity
  • Availability
  • Add to Cart