ENA196830289 Enamine

7-chloro-5-(trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD09041875
  • Cas Number : 926230-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45199
Availability details for
Quantity
  • Availability
  • Add to Cart