ENA196830291 Enamine

1-(benzenesulfonyl)-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD09045729
  • Cas Number : 16257-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45204
Availability details for
Quantity
  • Availability
  • Add to Cart