ENA196830295 Enamine

2-[(3-chlorophenyl)amino]acetic acid

  • MDL number MFCD02255622
  • Cas Number : 10242-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45214
Availability details for
Quantity
  • Availability
  • Add to Cart