ENA196830296 Enamine

2-(4-propylpiperazin-1-yl)benzaldehyde

  • MDL number MFCD11156404
  • Cas Number : 1021240-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43770
Availability details for
Quantity
  • Availability
  • Add to Cart