ENA196830303 Enamine

1-propanoylpiperidine-3-carboxylic acid

  • MDL number MFCD09050530
  • Cas Number : 926246-53-9
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45237
Availability details for
Quantity
  • Availability
  • Add to Cart