ENA196830308 Enamine

4-[(carbamoylmethyl)amino]benzoic acid

  • MDL number MFCD12197178
  • Cas Number : 86364-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45264

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45264
Availability details for
Quantity
  • Availability
  • Add to Cart