ENA196830325 Enamine

2-(3-bromophenoxy)benzoic acid

  • MDL number MFCD11156621
  • Cas Number : 1021245-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45315
Availability details for
Quantity
  • Availability
  • Add to Cart