ENA196830330 Enamine

N-ethyl-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD08060121
  • Cas Number : 909191-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45328
Availability details for
Quantity
  • Availability
  • Add to Cart