ENA196830334 Enamine

4-amino-N-(2-methylpropyl)benzamide

  • MDL number MFCD09043727
  • Cas Number : 38681-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45386
Availability details for
Quantity
  • Availability
  • Add to Cart