ENA196830338 Enamine

2-methanesulfonamido-3-methylbutanoic acid

  • MDL number MFCD06434618
  • Cas Number : 189945-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45398

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45398
Availability details for
Quantity
  • Availability
  • Add to Cart