ENA196830344 Enamine

N-(4-methyl-1,3-thiazol-2-yl)piperidine-4-carboxamide

  • MDL number MFCD09802917
  • Cas Number : 1016703-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45425
Availability details for
Quantity
  • Availability
  • Add to Cart