ENA196830348 Enamine

1-(3-aminophenyl)-5-ethyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD12197025
  • Cas Number : 1147675-34-0
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43815
Availability details for
Quantity
  • Availability
  • Add to Cart