ENA196830349 Enamine

1-[6-(2,6-dimethoxyphenoxy)pyridin-3-yl]methanamine

  • MDL number MFCD09732685
  • Cas Number : 953897-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45454
Availability details for
Quantity
  • Availability
  • Add to Cart