ENA196830351 Enamine

2-(3-amino-4-hydroxyphenyl)-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD12197028
  • Cas Number : 1147800-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43823
Availability details for
Quantity
  • Availability
  • Add to Cart