ENA196830353 Enamine

4-fluoro-2-(methylsulfanyl)benzoic acid

  • MDL number MFCD08444546
  • Cas Number : 410545-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43825
Availability details for
Quantity
  • Availability
  • Add to Cart