ENA238710064 Enamine

(5E)-5-{[4-(difluoromethoxy)-3-methoxyphenyl]methylidene}-1,3-thiazolidine-2,4-dione

  • MDL number MFCD11857757
  • Cas Number : 894259-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15633
Availability details for
Quantity
  • Availability
  • Add to Cart