ENA238710068 Enamine

2-amino-1-(4-ethylpiperazin-1-yl)-3-phenylpropan-1-one dihydrochloride

  • MDL number MFCD12912727
  • Cas Number : 1214636-82-4
  • Molecular Weight : 334.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-30210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30210
Availability details for
Quantity
  • Availability
  • Add to Cart