ENA238710072 Enamine

2-chloro-N-(6-methoxypyridin-3-yl)acetamide hydrochloride

  • MDL number MFCD12912731
  • Cas Number : 123578-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34063

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34063
Availability details for
Quantity
  • Availability
  • Add to Cart