ENA238710073 Enamine

2-(4-methyl-1,3-thiazol-2-yl)-1-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD12912732
  • Cas Number : 1193387-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34454
Availability details for
Quantity
  • Availability
  • Add to Cart