ENA238710082 Enamine

[4-(methanesulfonylmethyl)phenyl]methanamine

  • MDL number MFCD12912743
  • Cas Number : 854304-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41349
Availability details for
Quantity
  • Availability
  • Add to Cart