ENA238710085 Enamine

2-{2H,4H,5H,6H-cyclopenta[c]pyrazol-3-yl}aniline

  • MDL number MFCD12197001
  • Cas Number : 1181458-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41821

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41821
Availability details for
Quantity
  • Availability
  • Add to Cart